Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324423.1 | NP_001311352.1 | 4 | 673 | ||
NM_004664.3 | NP_004655.1 | 5 | 792 | |||
NR_136887.1 | 5 | 750 | ||||
NR_136888.1 | 6 | 869 | ||||
GenBank mRNA | AF028826.1 | - | 4 | 508 | ||
AF087693.1 | - | 5 | 761 | |||
AF173081.1 | - | 5 | 792 | |||
AK315321.1 | - | 5 | 636 | |||
BC062332.1 | - | 6 | 734 | |||
BC099921.1 | - | 5 | 602 | 6729473 | ||
BC118609.1 | - | 5 | 736 | 40109202 | ||
BC122561.1 | - | 5 | 736 | 40109204 | ||
CR407680.1 | - | 6 | 599 |