Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gimd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113782.1 | NP_001107254.1 | 2 | 488 | ||
XM_006233317.2 | XP_006233379.1 | 2 | 496 | |||
XM_008761516.1 | XP_008759738.1 | 2 | 288 | |||
GenBank mRNA | BC158622.1 | - | 2 | 488 | 7454123 |