Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108572.1 | NP_001102042.1 | 25 | 3521 | ||
XM_006233618.2 | XP_006233680.2 | 26 | 3637 | |||
XM_006233619.2 | XP_006233681.2 | 26 | 3628 | |||
XM_008761598.1 | XP_008759820.1 | 26 | 3979 | |||
XM_008761599.1 | XP_008759821.1 | 26 | 3976 | |||
XM_008761600.1 | XP_008759822.1 | 26 | 3958 | |||
XM_008761601.1 | XP_008759823.1 | 25 | 3835 | |||
XM_008761602.1 | XP_008759824.1 | 25 | 3814 |