Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypd6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134580.1 | NP_001128052.1 | 9 | 848 | ||
XM_008761762.1 | XP_008759984.1 | 6 | 2694 | |||
XM_008761763.1 | XP_008759985.1 | 7 | 3344 | |||
XM_008761764.1 | XP_008759986.1 | 6 | 3160 | |||
XM_008761765.1 | XP_008759987.1 | 8 | 3464 | |||
XM_008761766.1 | XP_008759988.1 | 5 | 2634 | |||
XM_008761767.1 | XP_008759989.1 | 5 | 3100 | |||
XM_008761768.1 | XP_008759990.1 | 5 | 2144 | |||
XM_008761769.1 | XP_008759991.1 | 6 | 2600 |