Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003878.2 | NP_003869.1 | 7 | 963 | ||
XM_011517623.2 | XP_011515925.1 | 7 | 714 | |||
GenBank mRNA | AK290450.1 | - | 7 | 707 | ||
AK301094.1 | - | 6 | 795 | |||
BC025025.1 | - | 7 | 764 | 3924620 | ||
U55206.1 | - | 7 | 739 |