Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014183.1 | NP_001014205.1 | 1 | 343 | ||
GenBank mRNA | BC087680.1 | - | 1 | 343 | 7318802 |