Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108596.1 | NP_001102066.1 | 9 | 790 | ||
XM_006235156.2 | XP_006235218.1 | 9 | 1272 |