Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003883.3 | NP_003874.2 | 12, 13 | 1037 | ||
XM_011537697.2 | XP_011535999.1 | 11, 12 | 889 | |||
GenBank mRNA | AF005482.1 | - | 10, 11 | 1053 | ||
AF039703.1 | - | 12, 13 | 993 | |||
BC000614.1 | - | 12, 13 | 1026 | 3345755 | ||
U66914.1 | - | 12, 13 | 982 | |||
U75696.1 | - | 11, 12 | 1020 | |||
U75697.1 | - | 12, 13 | 1026 |