Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108600.1 | NP_001102070.1 | 8 | 1675 | ||
XM_006235422.2 | XP_006235484.1 | 8 | 1725 |