Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014184.1 | NP_001014206.1 | 6 | 791 | ||
XM_006235487.2 | XP_006235549.1 | 8 | 2216 | |||
XM_006235489.2 | XP_006235551.1 | 8 | 1567 | |||
XM_006235490.2 | XP_006235552.1 | 9 | 2361 | |||
XM_006235491.2 | XP_006235553.1 | 8 | 1827 | |||
XM_006235493.2 | XP_006235555.1 | 7 | 1361 | |||
XM_006235494.2 | XP_006235556.1 | 7 | 2010 | |||
GenBank mRNA | BC089878.1 | - | 6 | 791 | 7326188 |