Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PROM1 SIRNA Assays ›
Gene Symbol:
PROM1
Gene Name:
prominin 1
Gene Aliases:
AC133, CD133, CORD12, MCDR2, MSTP061, PROML1, RP41, STGD4
Chromosome Location:
Chr.4: 15968226 - 16084100 on Build GRCh38
UniGene ID:
Hs.614734
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16880
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PROM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145847.1
NP_001139319.1
4
896
NM_001145848.1
NP_001139320.1
4
612
NM_001145849.1
NP_001139321.1
4
616
NM_001145850.1
NP_001139322.1
4
616
NM_001145851.1
NP_001139323.1
3
589
NM_001145852.1
NP_001139324.1
3
589
NM_006017.2
NP_006008.1
4
616
XM_005248195.4
XP_005248252.1
4
712
XM_005248196.4
XP_005248253.1
4
712
XM_006713974.3
XP_006714037.1
3
217
XM_011513890.1
XP_011512192.1
5
698
XM_011513892.2
XP_011512194.1
5
849
XM_011513893.2
XP_011512195.1
5
923
XM_011513894.2
XP_011512196.1
5
1008
XM_011513895.2
XP_011512197.1
6
1124
XM_011513896.2
XP_011512198.1
5
753
XM_011513897.2
XP_011512199.1
5
940
XM_011513898.2
XP_011512200.2
5
698
XM_011513899.2
XP_011512201.2
4
712
XM_011513900.2
XP_011512202.1
5
698
XM_011513902.2
XP_011512204.1
5
698
XM_011513903.2
XP_011512205.1
4
247
XM_017008799.1
XP_016864288.1
4
742
XM_017008800.1
XP_016864289.1
5
698
XM_017008801.1
XP_016864290.1
5
698
XM_017008802.1
XP_016864291.1
5
790
XM_017008803.1
XP_016864292.1
4
687
XM_017008804.1
XP_016864293.1
4
757
XM_017008805.1
XP_016864294.1
4
630
GenBank mRNA
AF027208.1
-
4
441
AF117225.1
-
4
657
AF507034.1
-
3
414
AK027420.1
-
4
635
AK027422.1
-
4
657
AK075411.1
-
4
612
AK226033.1
-
4
635
AK310415.1
-
4
462
AK314529.1
-
4
657
AK315889.1
-
1
45
AY449689.1
-
3
377
AY449690.1
-
3
377
AY449691.1
-
3
377
AY449692.1
-
4
404
AY449693.1
-
4
404
BC012089.1
-
4
612
4644690
HQ628626.1
-
3
377
HQ628627.1
-
5
633
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
retina layer formation
photoreceptor cell maintenance
retina morphogenesis in camera-type eye
camera-type eye photoreceptor cell differentiation
glomerular visceral epithelial cell differentiation
glomerular parietal epithelial cell differentiation
positive regulation of nephron tubule epithelial cell differentiation
protein binding
actinin binding
cadherin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline