Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbpjl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108604.1 | NP_001102074.1 | 11, 12 | 1419 | ||
XM_006235598.2 | XP_006235660.1 | 10, 11 | 1361 |