Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102555457 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235631.2 | XP_006235693.1 | 16 | 1572 |
Gene Symbol: Elmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134955.1 | NP_001128427.1 | 17 | 1596 | ||
XM_006235599.2 | XP_006235661.1 | 16 | 1572 |