Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135801.1 | NP_001129273.1 | 20 | 2232 | ||
GenBank mRNA | BC088338.1 | - | 8 | 747 | ||
BC160885.1 | - | 20 | 2232 | 7384442 |