Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCAR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006018.2 | NP_006009.2 | 1 | 567 | ||
GenBank mRNA | AK290930.1 | - | 1 | 448 | ||
AK313212.1 | - | 1 | 466 | |||
BC038955.1 | - | 1 | 457 | |||
BC047891.1 | - | 1 | 472 | 5750043 | ||
BI908162.1 | - | 1 | 459 | |||
CU451507.1 | - | 1 | 109 | |||
D10923.1 | - | 1 | 472 | |||
DA953971.1 | - | 1 | 486 |
Gene Symbol: HCAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177551.3 | NP_808219.1 | 1 | 486 | ||
GenBank mRNA | AK290915.1 | - | 1 | 425 | ||
AL698846.1 | - | 1 | 486 | |||
AY148884.1 | - | 1 | 472 | |||
BC027965.1 | - | 1 | 472 | 5222688 | ||
BC056419.1 | - | 1 | 458 | 5747743 |