Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113752.1 | NP_001107224.1 | 5 | 1577 | ||
GenBank mRNA | BC158660.1 | - | 5 | 1577 | 8369614 |