Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108616.1 | NP_001102086.1 | 7 | 1745 | ||
XM_006235938.2 | XP_006236000.1 | 10 | 1855 | |||
XM_006235939.2 | XP_006236001.1 | 10 | 1854 | |||
XM_006235940.2 | XP_006236002.1 | 10 | 1853 | |||
XM_006235941.2 | XP_006236003.1 | 10 | 1544 | |||
XM_006235942.2 | XP_006236004.1 | 10 | 1543 | |||
XM_008762650.1 | XP_008760872.1 | 10 | 1159 | |||
GenBank mRNA | KF447874.1 | - | 10 | 1249 |