Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243797.1 | NP_001230726.1 | 2, 3 | 127 | ||
NM_001243798.1 | NP_001230727.1 | 2, 3 | 132 | |||
NM_006022.3 | NP_006013.1 | 2, 3 | 469 | |||
NM_183422.3 | NP_904358.2 | 2, 3 | 3451 | |||
GenBank mRNA | AB082525.1 | - | 2, 3 | 3451 | ||
AJ222700.1 | - | 2, 3 | 364 | |||
AK027071.1 | - | 2, 3 | 2006 | |||
AK290751.1 | - | 2, 3 | 404 | |||
AK312345.1 | - | 2, 3 | 403 | |||
BC000456.2 | - | 2, 3 | 385 | |||
BC016867.1 | - | 2, 3 | 388 | |||
BC069207.1 | - | 2, 3 | 377 | |||
BC105277.1 | - | 2, 3 | 380 | |||
BC136475.1 | - | 2, 3 | 3221 | 9020464 | ||
BC136482.1 | - | 2, 3 | 3221 | 9020471 | ||
BC146664.1 | - | 2, 3 | 384 | |||
BC146679.1 | - | 2, 3 | 384 | |||
BF307401.1 | - | 2, 3 | 129 | |||
BG703493.1 | - | 2, 3 | 475 | |||
BI463370.1 | - | 2, 3 | 135 | |||
CN344671.1 | - | 2, 3 | 107 | |||
CR456711.1 | - | 2, 3 | 173 | |||
CR627459.1 | - | 3, 4 | 1644 | |||
D38585.1 | - | 2, 3 | 173 | |||
D87061.1 | - | 2, 3 | 173 | |||
U35048.1 | - | 2, 3 | 355 | |||
X97299.1 | - | 1, 2 | 57 |