Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108620.1 | NP_001102090.1 | 2 | 169 | ||
XM_008762795.1 | XP_008761017.1 | 2 | 346 | |||
XM_008762796.1 | XP_008761018.1 | 2 | 347 |