Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012729.1 | NP_036861.1 | 3 | 235 | ||
GenBank mRNA | V01274.1 | - | 3 | 212 |
Gene Symbol: LOC103690254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762925.1 | XP_008761147.1 | 3 | 235 |
Gene Symbol: LOC683849 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749732.3 | XP_003749780.1 | 3 | 235 |
Gene Symbol: Prss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012635.1 | NP_036767.1 | 3 | 247 | ||
GenBank mRNA | FQ211333.1 | - | 3 | 250 | ||
FQ226369.1 | - | 3 | 248 | |||
FQ227841.1 | - | 3 | 248 | |||
FQ233648.1 | - | 3 | 248 | |||
FQ235029.1 | - | 3 | 248 | |||
V01273.1 | - | 3 | 247 |
Gene Symbol: Try10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004097.1 | NP_001004097.1 | 3 | 235 | ||
XM_008762901.1 | XP_008761123.1 | 1 | 55 |
Gene Symbol: Try4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108626.1 | NP_001102096.1 | 3 | 252 | ||
GenBank mRNA | FQ231537.1 | - | 3 | 249 |