Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318787.1 | NP_001305716.1 | 4 | 805 | ||
NM_001318789.1 | NP_001305718.1 | 3 | 595 | |||
NM_001318790.1 | NP_001305719.1 | 3 | 572 | |||
NM_001318791.1 | NP_001305720.1 | 3 | 561 | |||
NM_001318793.1 | NP_001305722.1 | 3 | 557 | |||
NM_001318795.1 | NP_001305724.1 | 3 | 536 | |||
NM_001318796.1 | NP_001305725.1 | 3 | 521 | |||
NM_003264.4 | NP_003255.2 | 3 | 580 | |||
XM_011532215.2 | XP_011530517.1 | 3 | 505 | |||
XM_011532216.2 | XP_011530518.1 | 2 | 452 | |||
XM_017008573.1 | XP_016864062.1 | 4 | 1051 | |||
XM_017008574.1 | XP_016864063.1 | 4 | 733 | |||
XM_017008575.1 | XP_016864064.1 | 4 | 777 | |||
XM_017008576.1 | XP_016864065.1 | 2 | 1351 | |||
GenBank mRNA | AB445624.1 | - | 1 | 324 | ||
AF051152.1 | - | 2 | 486 | |||
AF502291.1 | - | 1 | 324 | |||
AK125659.1 | - | 1 | 1728 | |||
BC033756.1 | - | 3 | 474 | 5213439 | ||
BI769109.1 | - | 3 | 543 | |||
DA460298.1 | - | 4 | 546 | |||
DA870971.1 | - | 4 | 561 | |||
DQ012265.1 | - | 1 | 324 | |||
DQ012266.1 | - | 1 | 324 | |||
DQ012267.1 | - | 1 | 324 | |||
DQ012268.1 | - | 1 | 324 | |||
DQ012269.1 | - | 1 | 324 | |||
DQ012270.1 | - | 1 | 324 | |||
DQ012271.1 | - | 1 | 324 | |||
KU178364.1 | - | 1 | 324 | |||
U88878.1 | - | 2 | 453 |