Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100749.1 | NP_001094219.1 | 4, 5 | 450 | ||
GenBank mRNA | AY328517.1 | - | 3, 4 | 229 | ||
AY386318.1 | - | 4, 5 | 292 |