Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2cd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134581.1 | NP_001128053.1 | 2, 3 | 313 | ||
XM_006237633.2 | XP_006237695.1 | 2, 3 | 343 | |||
XM_006237634.2 | XP_006237696.1 | 2, 3 | 344 | |||
XM_006237636.2 | XP_006237698.1 | 2, 3 | 341 | |||
XM_006237637.2 | XP_006237699.1 | 2, 3 | 343 | |||
XM_006237638.2 | XP_006237700.1 | 2, 3 | 341 | |||
XM_006237639.2 | XP_006237701.1 | 2, 3 | 340 | |||
XM_006237640.2 | XP_006237702.1 | 2, 3 | 341 | |||
XM_008763400.1 | XP_008761622.1 | 2, 3 | 342 | |||
XM_008763401.1 | XP_008761623.1 | 2, 3 | 341 | |||
GenBank mRNA | FQ231190.1 | - | 2, 3 | 238 |