Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cspp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191864.1 | NP_001178793.1 | 20 | 2446 | ||
XM_006237763.2 | XP_006237825.1 | 21 | 2742 | |||
XM_006237767.2 | XP_006237829.1 | 20 | 2589 | |||
XM_006237774.2 | XP_006237836.1 | 16 | 2178 | |||
XM_006237775.2 | XP_006237837.1 | 21 | 2739 | |||
XM_006237776.2 | XP_006237838.1 | 20 | 2715 | |||
XM_006237777.2 | XP_006237839.1 | 19 | 2634 | |||
XM_006237778.2 | XP_006237840.1 | 20 | 2586 | |||
XM_008763498.1 | XP_008761720.1 | 21 | 2742 | |||
XM_008763499.1 | XP_008761721.1 | 21 | 2730 | |||
XM_008763500.1 | XP_008761722.1 | 20 | 2718 | |||
XM_008763501.1 | XP_008761723.1 | 19 | 2637 | |||
XM_008763502.1 | XP_008761724.1 | 21 | 2742 | |||
XM_008763503.1 | XP_008761725.1 | 18 | 2538 | |||
XM_008763504.1 | XP_008761726.1 | 21 | 2653 | |||
XM_008763505.1 | XP_008761727.1 | 20 | 2538 | |||
XM_008763506.1 | XP_008761728.1 | 21 | 3229 | |||
GenBank mRNA | BC166546.1 | - | 2 | 264 |