Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PER3 SIRNA Assays ›
Gene Symbol:
PER3
Gene Name:
period circadian clock 3
Gene Aliases:
FASPS3, GIG13
Chromosome Location:
Chr.1: 7784285 - 7845181 on Build GRCh38
UniGene ID:
Hs.162200
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16927
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PER3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289861.1
NP_001276790.1
5
727
NM_001289862.1
NP_001276791.1
5
727
NM_001289863.1
NP_001276792.1
5
727
NM_001289864.1
NP_001276793.1
5
727
NM_016831.2
NP_058515.1
5
727
XM_005263524.3
XP_005263581.1
4
602
XM_011542385.2
XP_011540687.1
4
602
XM_011542390.2
XP_011540692.1
4
602
XM_017002723.1
XP_016858212.1
4
602
XM_017002724.1
XP_016858213.1
4
602
XM_017002725.1
XP_016858214.1
4
602
XM_017002726.1
XP_016858215.1
4
602
XM_017002727.1
XP_016858216.1
6
921
XM_017002728.1
XP_016858217.1
4
663
XM_017002729.1
XP_016858218.1
2
106
XM_017002730.1
XP_016858219.1
4
663
XM_017002731.1
XP_016858220.1
4
663
XM_017002732.1
XP_016858221.1
3
712
XM_017002733.1
XP_016858222.1
6
828
XM_017002734.1
XP_016858223.1
7
970
XM_017002735.1
XP_016858224.1
6
828
XM_017002736.1
XP_016858225.1
7
970
XM_017002737.1
XP_016858226.1
2
213
XM_017002738.1
XP_016858227.1
2
219
GenBank mRNA
AB047686.1
-
4
687
AK307760.1
-
4
602
AK308303.1
-
5
776
AL157954.1
-
4
638
AY493418.1
-
4
534
BC026102.1
-
5
749
BC144137.1
-
4
631
BC146781.1
-
4
687
9007235
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
circadian regulation of gene expression
regulation of circadian sleep/wake cycle, sleep
protein stabilization
protein binding
kinase binding
ubiquitin protein ligase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0