Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173433.1 | NP_001166904.1 | 9 | 805 | ||
XM_006238281.2 | XP_006238343.1 | 9 | 1619 | |||
XM_006238283.2 | XP_006238345.1 | 9 | 1588 | |||
XM_006238284.2 | XP_006238346.1 | 9 | 1586 | |||
XM_006238286.2 | XP_006238348.1 | 9 | 1618 | |||
XM_006238287.2 | XP_006238349.1 | 9 | 1588 | |||
XM_006238288.2 | XP_006238350.1 | 9 | 1585 | |||
XM_006238289.2 | XP_006238351.1 | 8 | 657 | |||
XM_006238290.2 | XP_006238352.1 | 8 | 1802 | |||
XM_006238293.2 | XP_006238355.1 | 9 | 1585 | |||
XM_008763785.1 | XP_008762007.1 | 8 | 808 | |||
XM_008763786.1 | XP_008762008.1 | 8 | 805 | |||
XM_008763787.1 | XP_008762009.1 | 8 | 777 | |||
XM_008763788.1 | XP_008762010.1 | 8 | 775 | |||
XM_008763789.1 | XP_008762011.1 | 8 | 807 | |||
XM_008763790.1 | XP_008762012.1 | 8 | 807 | |||
XM_008763791.1 | XP_008762013.1 | 6 | 1840 | |||
XM_008763792.1 | XP_008762014.1 | 6 | 1336 | |||
XM_008763793.1 | XP_008762015.1 | 7 | 641 |