Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108670.1 | NP_001102140.1 | 3 | 634 | ||
XM_006238417.2 | XP_006238479.1 | 3 | 516 | |||
XM_006238418.2 | XP_006238480.1 | 3 | 516 | |||
XM_006238419.2 | XP_006238481.1 | 2 | 411 |