Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Raver2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191867.1 | NP_001178796.1 | 8 | 1408 | ||
XM_006238451.2 | XP_006238513.1 | 8 | 1466 |