Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160302.1 | NP_001153774.1 | 14 | 1720 | ||
NM_001160306.1 | NP_001153778.1 | 13 | 1705 | |||
NM_003895.3 | NP_003886.3 | 14 | 1712 | |||
NM_203446.2 | NP_982271.2 | 14 | 1712 | |||
XM_017028494.1 | XP_016883983.1 | 14 | 1702 | |||
XM_017028495.1 | XP_016883984.1 | 13 | 1712 | |||
XM_017028496.1 | XP_016883985.1 | 14 | 1702 | |||
XM_017028497.1 | XP_016883986.1 | 14 | 1800 | |||
XM_017028498.1 | XP_016883987.1 | 14 | 1702 | |||
XM_017028499.1 | XP_016883988.1 | 13 | 1712 | |||
XM_017028500.1 | XP_016883989.1 | 14 | 1702 | |||
XM_017028501.1 | XP_016883990.1 | 14 | 1702 | |||
XM_017028502.1 | XP_016883991.1 | 14 | 1702 | |||
XM_017028503.1 | XP_016883992.1 | 14 | 1702 | |||
XM_017028504.1 | XP_016883993.1 | 14 | 1702 | |||
XM_017028505.1 | XP_016883994.1 | 13 | 1712 | |||
GenBank mRNA | AB020717.2 | - | 14 | 1704 | ||
AF009039.1 | - | 14 | 1638 | |||
AF009040.1 | - | 14 | 1638 | |||
AK307762.1 | - | 14 | 1702 | |||
BC019630.1 | - | 13 | 1687 | |||
BC098395.1 | - | 13 | 1666 | |||
BC136603.1 | - | 12 | 1601 | 9020593 |