Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009710.1 | NP_001009710.1 | 2 | 254 | ||
XM_006238682.2 | XP_006238744.1 | 2 | 639 | |||
GenBank mRNA | BC087037.1 | - | 2 | 254 | 7133544 |