Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030052.1 | NP_001025223.1 | 2 | 148 | ||
XM_006238827.2 | XP_006238889.1 | 2 | 161 | |||
XM_006238828.2 | XP_006238890.1 | 2 | 122 | |||
GenBank mRNA | BC168721.1 | - | 2 | 126 | 6920432 | |
FQ224355.1 | - | 2 | 106 |
Gene Symbol: LOC100363268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729080.3 | XP_002729126.1 | 1 | 103 |