Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003897.3 | NP_003888.2 | 2 | 758 | ||
GenBank mRNA | AF039067.1 | - | 1 | 835 | ||
AK222588.1 | - | 2 | 755 | |||
BC000844.2 | - | 2 | 742 | 3457670 | ||
BC005080.1 | - | 2 | 744 | 3617088 | ||
BM915952.1 | - | 2 | 763 | |||
S81914.1 | - | 2 | 742 | |||
Y14551.1 | - | 2 | 737 |