Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3d21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162535.1 | NP_001156007.1 | 10 | 870 | ||
XM_008764049.1 | XP_008762271.1 | 13 | 2029 | |||
GenBank mRNA | BC162054.1 | - | 10 | 870 |