Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGEF7 SIRNA Assays ›
Gene Symbol:
ARHGEF7
Gene Name:
Rho guanine nucleotide exchange factor 7
Gene Aliases:
BETA-PIX, COOL-1, COOL1, Nbla10314, P50, P50BP, P85, P85COOL1, P85SPR, PAK3, PIXB
Chromosome Location:
Chr.13: 111114619 - 111305734 on Build GRCh38
UniGene ID:
Hs.508738
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16948
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGEF7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113511.2
NP_001106983.1
15
2496
NM_001113512.2
NP_001106984.1
13
2346
NM_001113513.1
NP_001106985.1
13
1438
NM_001320851.1
NP_001307780.1
12
1219
NM_001320852.1
NP_001307781.1
14
2433
NM_001320853.1
NP_001307782.1
13
1344
NM_001320854.1
NP_001307783.1
9
958
NM_003899.3
NP_003890.1
13
1551
NM_145735.3
NP_663788.1
14
2433
XM_006719956.3
XP_006720019.1
14
1561
XM_017020814.1
XP_016876303.1
14
1561
XM_017020815.1
XP_016876304.1
12
1554
XM_017020816.1
XP_016876305.1
13
1329
XM_017020817.1
XP_016876306.1
12
1554
XM_017020818.1
XP_016876307.1
14
1429
XM_017020819.1
XP_016876308.1
13
1438
XM_017020820.1
XP_016876309.1
13
1561
XM_017020821.1
XP_016876310.1
13
1256
XM_017020822.1
XP_016876311.1
12
1252
XM_017020823.1
XP_016876312.1
12
2029
XM_017020824.1
XP_016876313.1
14
1429
XM_017020825.1
XP_016876314.1
12
1180
XM_017020826.1
XP_016876315.1
14
1429
XM_017020827.1
XP_016876316.1
12
1253
XM_017020828.1
XP_016876317.1
10
877
XM_017020829.1
XP_016876318.1
10
877
GenBank mRNA
AB075521.1
-
4
292
AB177848.1
-
14
2434
AB177849.1
-
11
1157
AK295472.1
-
9
883
AK300272.1
-
13
1331
AL834228.1
-
14
1617
BC033905.1
-
13
1543
5262912
BC050521.1
-
13
1410
BC060776.1
-
15
1838
5271977
D63476.1
-
13
1527
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
hematopoietic progenitor cell differentiation
Golgi organization
signal transduction
nervous system development
positive regulation of fibroblast migration
lamellipodium assembly
positive regulation of protein binding
regulation of Rho protein signal transduction
intracellular signal transduction
negative regulation of epidermal growth factor receptor signaling pathway
positive regulation of apoptotic process
positive regulation of GTPase activity
ephrin receptor signaling pathway
focal adhesion assembly
regulation of small GTPase mediated signal transduction
positive regulation of growth hormone secretion
positive regulation of substrate adhesion-dependent cell spreading
regulation of GTP binding
positive regulation of lamellipodium morphogenesis
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0