Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198785.1 | NP_942080.1 | 16, 17 | 2272 | ||
XM_006239234.2 | XP_006239296.1 | 17, 18 | 2306 | |||
XM_006239235.2 | XP_006239297.1 | 17, 18 | 2303 | |||
XM_006239236.2 | XP_006239298.1 | 17, 18 | 2303 | |||
XM_006239237.2 | XP_006239299.1 | 17, 18 | 2300 | |||
GenBank mRNA | AB073880.1 | - | 16, 17 | 2272 |
Gene Symbol: LOC100911993 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751808.3 | XP_003751856.1 | 1, 2 | 3076 | ||
XM_006254970.2 | XP_006255032.1 | 1, 2 | 3076 |