Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108692.1 | NP_001102162.1 | 5 | 992 | ||
XM_008764270.1 | XP_008762492.1 | 3 | 579 | |||
XM_008764271.1 | XP_008762493.1 | 3 | 539 | |||
XM_008764272.1 | XP_008762494.1 | 3 | 1818 |