Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108700.1 | NP_001102170.1 | 9, 10 | 969 | ||
XM_006239636.2 | XP_006239698.1 | 9, 10 | 1245 | |||
XM_008764424.1 | XP_008762646.1 | 10, 11 | 1086 | |||
GenBank mRNA | FQ225505.1 | - | 9, 10 | 977 |