Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem178a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004282.1 | NP_001004282.1 | 2, 3 | 526 | ||
GenBank mRNA | BC079338.1 | - | 2, 3 | 556 | 7106500 |