Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tssc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012192.1 | NP_001012192.1 | 6, 7 | 735 | ||
GenBank mRNA | BC087633.1 | - | 6, 7 | 735 | 7320156 |