Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108712.2 | NP_001102182.2 | 4 | 620 | ||
XM_006240120.2 | XP_006240182.1 | 4 | 638 | |||
GenBank mRNA | BC166982.1 | - | 4 | 547 |