Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005171.4 | NP_005162.1 | 3 | 442 | ||
XM_011538386.2 | XP_011536688.1 | 3 | 2499 | |||
XM_011538387.2 | XP_011536689.1 | 3 | 172 | |||
XM_011538388.2 | XP_011536690.1 | 3 | 373 | |||
XM_017019331.1 | XP_016874820.1 | 3 | 212 | |||
XM_017019332.1 | XP_016874821.1 | 3 | 306 | |||
XM_017019333.1 | XP_016874822.1 | 3 | 192 | |||
XM_017019334.1 | XP_016874823.1 | 3 | 218 | |||
XM_017019335.1 | XP_016874824.1 | 3 | 216 | |||
XM_017019336.1 | XP_016874825.1 | 3 | 211 | |||
GenBank mRNA | AK313406.1 | - | 3 | 347 | ||
BC029619.1 | - | 3 | 156 | 4102231 | ||
BG502726.1 | - | 3 | 175 | |||
BG545632.1 | - | 3 | 370 | |||
BI712653.1 | - | 3 | 331 | |||
DA468394.1 | - | 3 | 442 |