Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108715.1 | NP_001102185.1 | 5 | 1078 | ||
XM_006240286.2 | XP_006240348.1 | 5 | 1426 | |||
XM_006240287.2 | XP_006240349.1 | 5 | 1261 | |||
GenBank mRNA | BC166432.1 | - | 5 | 1242 | 7135396 |