Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100863.1 | NP_001094333.1 | 5 | 713 | ||
XM_006240288.2 | XP_006240350.1 | 6 | 1844 |