Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317086.1 | NP_001304015.1 | 4, 5 | 466 | ||
NM_003904.4 | NP_003895.1 | 5, 6 | 628 | |||
GenBank mRNA | AF019767.1 | - | 5, 6 | 608 | ||
AK301227.1 | - | 5, 6 | 576 | |||
BC004256.1 | - | 5, 6 | 583 | 3610932 | ||
BC012162.1 | - | 5, 6 | 596 | 4578136 | ||
BC017349.2 | - | 5, 6 | 613 | 5013820 | ||
BC017380.2 | - | 5, 6 | 596 | 2988688 | ||
BC111028.1 | - | 5, 6 | 600 | 5245870 | ||
BT006642.1 | - | 5, 6 | 580 | |||
CD359945.1 | - | 5, 6 | 628 | |||
CR975994.1 | - | 2 | 298 |
Gene Symbol: ZNF259P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR627448.1 | - | 1 | 563 |