Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alkbh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240394.2 | XP_006240456.1 | 5 | 586 | ||
GenBank mRNA | FQ225419.1 | - | 3 | 161 | ||
FQ230972.1 | - | 3 | 161 |