Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108719.2 | NP_001102189.1 | 5 | 494 | ||
XM_008764816.1 | XP_008763038.1 | 11 | 1204 | |||
GenBank mRNA | BC168996.1 | - | 5 | 494 | 7319639 | |
FQ210625.1 | - | 7 | 622 |