Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008776.2 | NP_001008776.1 | 1 | 166 | ||
NM_001166352.1 | NP_001159824.1 | 2 | 272 | |||
XM_006240481.2 | XP_006240543.1 | 2 | 271 | |||
GenBank mRNA | AY325135.1 | - | 1 | 206 | ||
BC111708.1 | - | 1 | 184 | 7368690 | ||
EX492846.1 | - | 2 | 272 | |||
FQ211426.1 | - | 2 | 244 |