Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021095.2 | NP_066918.2 | 7, 8 | 1231 | ||
NR_028323.1 | 7, 8 | 1231 | ||||
XM_006712128.1 | XP_006712191.1 | 7, 8 | 1026 | |||
XM_006712129.1 | XP_006712192.1 | 7, 8 | 1074 | |||
XM_006712130.1 | XP_006712193.1 | 7, 8 | 1065 | |||
XM_017005216.1 | XP_016860705.1 | 4, 5 | 801 | |||
GenBank mRNA | AF069307.1 | - | 7, 8 | 1127 | ||
AF081571.1 | - | 6, 7 | 943 | |||
AK027855.1 | - | 7, 8 | 1072 | |||
AK293162.1 | - | 1, 2 | 115 | |||
AK314545.1 | - | 8, 9 | 1058 | |||
BC012806.2 | - | 5, 6 | 793 | 3507776 | ||
BC015631.2 | - | 6, 7 | 944 | 3628126 |