Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021095.2 | NP_066918.2 | 3 | 779 | ||
NR_028323.1 | 3 | 779 | ||||
XM_006712128.1 | XP_006712191.1 | 3 | 574 | |||
XM_006712129.1 | XP_006712192.1 | 3 | 622 | |||
XM_006712130.1 | XP_006712193.1 | 3 | 613 | |||
XM_011533146.2 | XP_011531448.1 | 1 | 401 | |||
XM_017005216.1 | XP_016860705.1 | 1 | 401 | |||
GenBank mRNA | AF069307.1 | - | 3 | 675 | ||
AF081571.1 | - | 2 | 491 | |||
AK027855.1 | - | 3 | 620 | |||
AK314545.1 | - | 4 | 606 | |||
BC012806.2 | - | 1 | 341 | 3507776 | ||
BC015631.2 | - | 2 | 492 | 3628126 |