Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012195.1 | NP_001012195.1 | 3 | 282 | ||
XM_006240772.2 | XP_006240834.1 | 5 | 713 | |||
XM_006240773.2 | XP_006240835.1 | 5 | 775 | |||
XM_006240774.2 | XP_006240836.1 | 4 | 621 | |||
XM_006240775.2 | XP_006240837.1 | 5 | 631 | |||
XM_006240776.2 | XP_006240838.1 | 4 | 539 | |||
XM_008765030.1 | XP_008763252.1 | 4 | 1632 | |||
GenBank mRNA | BC083614.1 | - | 3 | 282 | 7133774 |